Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008290.1 | NP_001008291.1 | 1 - 2 | 333 | 104 | |
| GenBank mRNA | BC086384.1 | - | 1 - 2 | 333 | 7301624 | 104 |
| FQ220218.1 | - | 1 - 2 | 144 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086384.1, NM_001008290.1 transcripts.
Set Membership: |
|

Fam216a
Genomic Map