Assay Details
| Gene Symbol: Ccdc62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134766.1 | NP_001128238.1 | 6 - 7 | 896 | 102 | |
| XM_006249355.2 | XP_006249417.1 | 6 - 7 | 1764 | 102 | ||
| XM_006249356.2 | XP_006249418.1 | 6 - 7 | 1710 | 102 | ||
| XM_006249357.2 | XP_006249419.1 | 6 - 7 | 1764 | 102 | ||
| XM_006249358.1 | XP_006249420.1 | 6 - 7 | 837 | 102 | ||
| XM_008769265.1 | XP_008767487.1 | 5 - 6 | 649 | 102 | ||
| GenBank mRNA | BC086549.1 | - | 2 - 3 | 300 | 102 | |
| BC099139.1 | - | 2 - 3 | 175 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc62
Genomic Map