Assay Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124768.1 | NP_001118240.1 | 2 - 3 | 224 | 112 | |
| NM_001124769.1 | NP_001118241.1 | 2 - 3 | 224 | 112 | ||
| NM_001124770.1 | NP_001118242.1 | 2 - 3 | 224 | 112 | ||
| NM_012666.2 | NP_036798.1 | 2 - 3 | 224 | 112 | ||
| XM_008762692.1 | XP_008760914.1 | 2 - 3 | 365 | 112 | ||
| XM_008762693.1 | XP_008760915.1 | 2 - 3 | 362 | 112 | ||
| XM_008762694.1 | XP_008760916.1 | 2 - 3 | 368 | 112 | ||
| XM_008762695.1 | XP_008760917.1 | 2 - 3 | 366 | 112 | ||
| GenBank mRNA | M14312.1 | - | 1 - 2 | 136 | 112 | |
| M34183.1 | - | 2 - 3 | 226 | 112 | ||
| M34184.1 | - | 2 - 3 | 226 | 112 | ||
| X56306.1 | - | 1 - 2 | 131 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tac1
Genomic Map