Assay Details
| Gene Symbol: Ppp1r3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109222.1 | NP_001102692.1 | 3 - 4 | 1041 | 125 | |
| XM_008762717.1 | XP_008760939.1 | 3 - 4 | 1083 | 125 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P037711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P037716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30541 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037716 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P037716 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P037716 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P037711 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90019 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10861134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861133 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10861132 | - | - |
| Rat Genome 230 2.0 Array | 1397559_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1397559_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400017661.rn.1 | TC0400002481.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397559_at | - | - |
| Rat Expression Set 230 | 1397559_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861132 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21681099 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371289 | - | - |

Ppp1r3a
Genomic Map