Assay Details
| Gene Symbol: Hyal6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024320.1 | NP_001019491.1 | 1 - 2 | 286 | 84 | |
| XM_006236165.2 | XP_006236227.1 | 1 - 2 | 298 | 84 | ||
| GenBank mRNA | BC082016.1 | - | 1 - 2 | 286 | 7132110 | 84 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC082016.1, NM_001024320.1, XM_006236165.2 transcripts.
Set Membership: |
|

Hyal6
Genomic Map