Assay Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109225.1 | NP_001102695.1 | 1 - 2 | 120 | 135 | |
| XM_008762812.1 | XP_008761034.1 | 2 - 3 | 140 | 135 | ||
| XM_008762813.1 | XP_008761035.1 | 1 - 2 | 121 | 135 | ||
| XM_008762814.1 | XP_008761036.1 | 2 - 3 | 285 | 135 | ||
| XM_008762815.1 | XP_008761037.1 | 2 - 3 | 361 | 135 | ||
| XM_008762816.1 | XP_008761038.1 | 2 - 3 | 140 | 135 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001109225.1, XM_008762812.1, XM_008762813.1, XM_008762814.1, XM_008762815.1, XM_008762816.1 transcripts.
Set Membership: |
|

Zfp800
Genomic Map