Assay Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109225.1 | NP_001102695.1 | 2 - 3 | 223 | 178 | |
| XM_008762812.1 | XP_008761034.1 | 3 - 4 | 243 | 178 | ||
| XM_008762813.1 | XP_008761035.1 | 2 - 3 | 224 | 178 | ||
| XM_008762814.1 | XP_008761036.1 | 3 - 4 | 388 | 178 | ||
| XM_008762815.1 | XP_008761037.1 | 3 - 4 | 464 | 178 | ||
| XM_008762816.1 | XP_008761038.1 | 3 - 4 | 243 | 178 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp800
Genomic Map