Assay Details
| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106585.1 | NP_001100055.1 | 1 - 2 | 172 | 76 | |
| GenBank mRNA | BC167094.1 | - | 1 - 2 | 43 | 7462085 | 76 |
| FQ211862.1 | - | 138 | 76 | |||
| FQ212479.1 | - | 138 | 76 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC167094.1,NM_001106585.1 transcripts.
This assay is in the 5'UTR region of the BC167094.1,NM_001106585.1 transcripts.
Set Membership: |
|

Lsm8
Genomic Map