Assay Details
| Gene Symbol: Lrguk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106589.1 | NP_001100059.1 | 1 - 2 | 308 | 64 | |
| XM_006236253.2 | XP_006236315.1 | 1 - 2 | 319 | 64 | ||
| XM_006236255.2 | XP_006236317.1 | 1 - 2 | 319 | 64 | ||
| XM_006236256.2 | XP_006236318.1 | 1 - 2 | 319 | 64 | ||
| XM_008762766.1 | XP_008760988.1 | 1 - 2 | 319 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lrguk
Genomic Map