Assay Details
| Gene Symbol: Tspan33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109227.1 | NP_001102697.1 | 4 - 5 | 570 | 73 | |
| XM_006236227.2 | XP_006236289.1 | 4 - 5 | 556 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tspan33
Genomic Map