Assay Details
| Gene Symbol: Adck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107855.1 | NP_001101325.1 | 4 - 5 | 1683 | 99 | |
| XM_006236325.2 | XP_006236387.1 | 3 - 4 | 1560 | 99 | ||
| XM_006236326.2 | XP_006236388.1 | 3 - 4 | 1559 | 99 | ||
| XM_008762782.1 | XP_008761004.1 | 4 - 5 | 1706 | 99 | ||
| GenBank mRNA | BC161918.1 | - | 3 - 4 | 1471 | 8369971 | 99 |
More Information
Updated Mapping Information
Set Membership: |
|

Adck2
Genomic Map