Assay Details
| Gene Symbol: Parp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749716.3 | XP_003749764.1 | 8 - 9 | 1846 | 59 | |
| XM_006236346.2 | XP_006236408.1 | 8 - 9 | 1846 | 59 | ||
More Information
Set Membership: |
|

Parp12
Genomic Map