Assay Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024318.1 | NP_001019489.1 | 1 - 2 | 197 | 148 | |
| GenBank mRNA | AY577765.1 | - | 1 - 2 | 197 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Asb4
Genomic Map