Assay Details
| Gene Symbol: LOC100363521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110645.1 | 2 - 3 | 125 | 139 | ||
| GenBank mRNA | FM094276.1 | - | 2 - 3 | 120 | 139 | |
| FM119392.1 | - | 2 - 3 | 125 | 139 | ||
| FQ224908.1 | - | 2 - 3 | 128 | 139 | ||
| FQ229518.1 | - | 2 - 3 | 128 | 139 | ||
More Information
Set Membership: |
|

LOC100363521
Genomic Map