Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153628.2 | NP_705892.1 | 2 - 3 | 253 | 93 | |
| GenBank mRNA | AB022714.2 | - | 2 - 3 | 254 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec11c
Genomic Map