Assay Details
| Gene Symbol: Sec11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153628.2 | NP_705892.1 | 3 - 4 | 395 | 143 | |
| GenBank mRNA | AB022714.2 | - | 3 - 4 | 396 | 143 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sec11c
Genomic Map