Assay Details
| Gene Symbol: Rbm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170484.1 | NP_001163955.1 | 1 - 2 | 83 | 95 | |
| XM_008760103.1 | XP_008758325.1 | 1 - 2 | 844 | 95 | ||
| GenBank mRNA | FM086093.1 | - | 1 - 2 | 69 | 95 | |
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC086338.1 | - | 67 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC086338.1, NM_001170484.1, XM_008760103.1 transcripts.
Set Membership: |
|

Rbm4
Genomic Map