Assay Details
| Gene Symbol: Rbm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760102.1 | XP_008758324.1 | 2 - 3 | 1413 | 71 | |
| XM_008760103.1 | XP_008758325.1 | 2 - 3 | 1270 | 71 | ||
| GenBank mRNA | FM086093.1 | - | 2 - 3 | 495 | 71 | |
| Gene Symbol: Rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC086338.1 | - | 493 | 71 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rbm4
Genomic Map