Assay Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134375.3 | NP_599202.2 | 368 | 54 | ||
| GenBank mRNA | DQ631800.1 | - | 368 | 54 | ||
| Gene Symbol: LOC103690037 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230507.2 | XP_006230569.2 | 2 - 3 | 1529 | 54 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nlrp6
Genomic Map