Assay Details
| Gene Symbol: LOC100911692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230597.2 | XP_006230659.1 | 1 - 2 | 781 | 74 | |
| Gene Symbol: Cracr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230561.2 | XP_006230623.1 | 1 - 2 | 781 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230561.2, XM_006230597.2 transcripts.
Set Membership: |
|

LOC100911692
Genomic Map