Assay Details
| Gene Symbol: LOC100911692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230595.2 | XP_006230657.1 | 7 - 8 | 2325 | 73 | |
| XM_006230596.2 | XP_006230658.1 | 8 - 9 | 1689 | 73 | ||
| XM_006230597.2 | XP_006230659.1 | 7 - 8 | 1577 | 73 | ||
| Gene Symbol: Cracr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127541.1 | NP_001121013.1 | 952 | 73 | ||
| XM_006230559.2 | XP_006230621.1 | 7 - 8 | 2325 | 73 | ||
| XM_006230561.2 | XP_006230623.1 | 7 - 8 | 1577 | 73 | ||
| GenBank mRNA | BC158864.1 | - | 952 | 7936496 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911692
Genomic Map