Assay Details
| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271039.1 | NP_001257968.1 | 2 - 3 | 132 | 60 | |
| XM_006231045.2 | XP_006231107.1 | 2 - 3 | 957 | 60 | ||
| GenBank mRNA | BC168207.1 | - | 1 - 2 | 101 | 7131074 | 60 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC168207.1, XM_006231045.2 transcripts.
Set Membership: |
|

Tmem216
Genomic Map