Assay Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033237.1 | NP_150240.1 | 1 - 2 | 146 | 107 | |
| GenBank mRNA | J03624.1 | - | 1 - 2 | 146 | 107 | |
| M18102.1 | - | 1 - 2 | 126 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the J03624.1, M18102.1, NM_033237.1 transcripts.
Set Membership: |
|

Gal
Genomic Map