Assay Details
| Gene Symbol: Gal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033237.1 | NP_150240.1 | 4 - 5 | 368 | 92 | |
| GenBank mRNA | J03624.1 | - | 4 - 5 | 368 | 92 | |
| M18102.1 | - | 4 - 5 | 348 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gal
Genomic Map