Assay Details
| Gene Symbol: LOC100911814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749086.3 | XP_003749134.2 | 8 - 9 | 1388 | 91 | |
| Gene Symbol: Gldc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107583.1 | NP_001101053.1 | 7 - 8 | 1283 | 91 | |
| XM_006231210.2 | XP_006231272.1 | 8 - 9 | 1387 | 91 | ||
| XM_006231211.2 | XP_006231273.1 | 8 - 9 | 1388 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

LOC100911814
Genomic Map