Assay Details
| Gene Symbol: Ms4a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760273.1 | XP_008758495.1 | 2 - 3 | 2769 | 84 | |
| XM_008760275.1 | XP_008758497.1 | 1 - 2 | 4384 | 84 | ||
| XM_342027.5 | XP_342028.3 | 1 - 2 | 4384 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008760273.1, XM_008760275.1, XM_342027.5 transcripts.
Set Membership: |
|

Ms4a4a
Genomic Map