Assay Details
| Gene Symbol: Rom1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009690.1 | NP_001009690.1 | 1 - 2 | 810 | 79 | |
| GenBank mRNA | AY212508.1 | - | 1 - 2 | 609 | 79 | |
| BC087605.1 | - | 1 - 2 | 810 | 7312188 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rom1
Genomic Map