Assay Details
| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC090345.1 | - | 5 - 6 | 811 | 93 | |
| BC107927.1 | - | 866 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC090345.1 | - | 5 - 6 | 811 | 93 | |
| BC107927.1 | - | 866 | 93 | |||
Set Membership: |
|