Assay Details
| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054005.1 | NP_446457.1 | 7 - 8 | 1633 | 92 | |
| GenBank mRNA | AF022147.1 | - | 7 - 8 | 1633 | 92 | |
| AF167170.1 | - | 7 - 8 | 1399 | 92 | ||
| BC090345.1 | - | 7 - 8 | 1379 | 92 | ||
| BC107927.1 | - | 1434 | 92 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cuzd1
Genomic Map