Assay Details
| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728846.4 | XP_002728892.3 | 4 - 5 | 2767 | 71 | |
| XM_006230916.2 | XP_006230978.1 | 4 - 5 | 2767 | 71 | ||
More Information
Set Membership: |
|

Phemx
Genomic Map