Assay Details
| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728846.4 | XP_002728892.3 | 6 - 7 | 2958 | 77 | |
| XM_006230915.2 | XP_006230977.1 | 5 - 6 | 2883 | 77 | ||
| GenBank mRNA | BC158837.1 | - | 4 - 5 | 423 | 77 | |
More Information
Set Membership: |
|

Phemx
Genomic Map