Assay Details
| Gene Symbol: Phemx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728846.4 | XP_002728892.3 | 8 - 9 | 3128 | 93 | |
| XM_006230915.2 | XP_006230977.1 | 7 - 8 | 3053 | 93 | ||
| XM_006230916.2 | XP_006230978.1 | 7 - 8 | 3041 | 93 | ||
| GenBank mRNA | BC088266.1 | - | 5 - 6 | 419 | 93 | |
| BC158837.1 | - | 6 - 7 | 593 | 93 | ||
More Information
Set Membership: |
|

Phemx
Genomic Map