Assay Details
| Gene Symbol: Ttll12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135922.1 | NP_001129394.1 | 5 - 6 | 841 | 57 | |
| XM_008765684.1 | XP_008763906.1 | 5 - 6 | 975 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll12
Genomic Map