Assay Details
| Gene Symbol: Tomm22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212514.2 | NP_997679.1 | 1 - 2 | 134 | 68 | |
| XM_006241974.1 | XP_006242036.1 | 2 - 3 | 223 | 68 | ||
| GenBank mRNA | AB162854.1 | - | 1 - 2 | 121 | 68 | |
| BC098639.1 | - | 1 - 2 | 134 | 7324771 | 68 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tomm22
Genomic Map