Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145676.1 | NP_663709.1 | 1 - 2 | 49 | 71 | |
| GenBank mRNA | AF393211.1 | - | 1 - 2 | 49 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF393211.1, NM_145676.1 transcripts.
Set Membership: |
|

Cby1
Genomic Map