Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145676.1 | NP_663709.1 | 2 - 3 | 165 | 60 | |
| XM_006241967.2 | XP_006242029.1 | 3 - 4 | 60 | |||
| GenBank mRNA | AF393211.1 | - | 2 - 3 | 165 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cby1
Genomic Map