Assay Details
| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241910.2 | XP_006241972.1 | 10 - 11 | 2746 | 69 | |
| GenBank mRNA | FQ235212.1 | - | 3 - 4 | 797 | 69 | |
More Information
Set Membership: |
|

LOC102548778
Genomic Map