Assay Details
| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241910.2 | XP_006241972.1 | 5 - 6 | 1512 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548778| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241910.2 | XP_006241972.1 | 5 - 6 | 1512 | 137 | |
Set Membership: |
|