Assay Details
| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241910.2 | XP_006241972.1 | 7 - 8 | 1803 | 146 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102548778| Gene Symbol: LOC102548778 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241910.2 | XP_006241972.1 | 7 - 8 | 1803 | 146 | |
Set Membership: |
|