Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171981.1 | NP_001165452.1 | 8 - 9 | 688 | 64 | |
| XM_008765682.1 | XP_008763904.1 | 7 - 8 | 1753 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sh3bp1
Genomic Map