Assay Details
| Gene Symbol: Lyc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128494.1 | NP_001121966.1 | 2 - 3 | 559 | 58 | |
| GenBank mRNA | BC166737.1 | - | 2 - 3 | 559 | 7937244 | 58 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Lyc2
Genomic Map