Assay Details
| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030031.1 | NP_001025202.1 | 18 - 19 | 1810 | 82 | |
| XM_006241251.2 | XP_006241313.1 | 17 - 18 | 1747 | 82 | ||
| GenBank mRNA | BC099819.1 | - | 18 - 19 | 1810 | 7930840 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
leukotriene metabolic process
response to zinc ion
leukotriene biosynthetic process
peptide catabolic process
response to peptide hormone
Type I pneumocyte differentiation
aminopeptidase activity
epoxide hydrolase activity
leukotriene-A4 hydrolase activity
metallopeptidase activity
zinc ion binding
poly(A) RNA binding
proteolysis
leukotriene metabolic process
response to zinc ion
leukotriene biosynthetic process
peptide catabolic process
response to peptide hormone
Type I pneumocyte differentiation
aminopeptidase activity
epoxide hydrolase activity
leukotriene-A4 hydrolase activity
metallopeptidase activity
zinc ion binding
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P618538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160128 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P618538 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P618538 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P618538 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P618538 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100542 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48584 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 836 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10894892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894879 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894887 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894880 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10894878 | - | - |
| Rat Genome 230 2.0 Array | 1389010_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389010_at | - | - |
| Affymetrix HT Rat Focus Array | 1389010_at | - | - |
| Affymetrix HT Rat Focus Array | 1389010_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700004269.rn.1 | TC0700000512.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700004270.rn.1 | TC0700000512.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389010_at | - | - |
| Rat Expression Set 230 | 1389010_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10894878 | - | - |
| Rat Genome U34 Set | S87522_g_at | - | - |
| Rat Genome U34 Set | S87522_at | - | - |
| Rat Genome U34 Set | rc_AA964325_at | - | - |
| Rat Genome U34 Set | rc_AA858804_s_at | - | - |
| Rat Genome U34 Set | rc_AA858552_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21201538 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361937 | - | - |

Lta4h
Genomic Map