Assay Details
| Gene Symbol: Gpihbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130547.1 | NP_001124019.1 | 1 - 2 | 92 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpihbp1| Gene Symbol: Gpihbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130547.1 | NP_001124019.1 | 1 - 2 | 92 | 68 | |
Set Membership: |
|