Assay Details
| Gene Symbol: Bik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053704.1 | NP_446156.1 | 1 - 2 | 97 | 119 | |
| GenBank mRNA | AF372501.1 | - | 1 - 2 | 97 | 119 | |
| FQ225338.1 | - | 1 - 2 | 74 | 119 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF372501.1, NM_053704.1 transcripts.
Set Membership: |
|

Bik
Genomic Map