Assay Details
| Gene Symbol: Ndufa4l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271272.1 | NP_001258201.1 | 3 - 4 | 213 | 80 | |
| GenBank mRNA | FM120013.1 | - | 3 - 4 | 216 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufa4l2
Genomic Map