Assay Details
| Gene Symbol: Cyc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277194.1 | NP_001264123.1 | 1 - 2 | 161 | 59 | |
| GenBank mRNA | FN801980.1 | - | 1 - 2 | 126 | 59 | |
| FQ214725.1 | - | 1 - 2 | 141 | 59 | ||
| FQ216469.1 | - | 1 - 2 | 143 | 59 | ||
| FQ220496.1 | - | 1 - 2 | 138 | 59 | ||
| FQ220832.1 | - | 1 - 2 | 143 | 59 | ||
| FQ222191.1 | - | 1 - 2 | 143 | 59 | ||
| FQ222396.1 | - | 1 - 2 | 143 | 59 | ||
| FQ224965.1 | - | 1 - 2 | 140 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyc1
Genomic Map