Assay Details
| Gene Symbol: Cyc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277194.1 | NP_001264123.1 | 3 - 4 | 494 | 99 | |
| GenBank mRNA | BC099171.1 | - | 1 - 2 | 838 | 99 | |
| FN801980.1 | - | 3 - 4 | 459 | 99 | ||
| FQ214725.1 | - | 3 - 4 | 474 | 99 | ||
| FQ220496.1 | - | 3 - 4 | 471 | 99 | ||
| FQ220832.1 | - | 3 - 4 | 476 | 99 | ||
| FQ222191.1 | - | 3 - 4 | 476 | 99 | ||
| FQ224965.1 | - | 3 - 4 | 473 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyc1
Genomic Map