Assay Details
| Gene Symbol: Slc16a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031744.2 | NP_113932.2 | 2 - 3 | 197 | 66 | |
| GenBank mRNA | AF059258.1 | - | 2 - 3 | 197 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF059258.1, NM_031744.2 transcripts.
Set Membership: |
|

Slc16a8
Genomic Map