Assay Details
| Gene Symbol: Cyb5r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138877.1 | NP_620232.1 | 3 - 4 | 268 | 74 | |
| XM_006242065.2 | XP_006242127.1 | 3 - 4 | 296 | 74 | ||
| GenBank mRNA | BC062066.1 | - | 3 - 4 | 284 | 5599255 | 74 |
| D00636.1 | - | 3 - 4 | 268 | 74 | ||
| J03867.1 | - | 3 - 4 | 263 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r3
Genomic Map