Assay Details
| Gene Symbol: Mars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127659.1 | NP_001121131.1 | 4 - 5 | 421 | 70 | |
| XM_006241451.2 | XP_006241513.1 | 4 - 5 | 534 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mars
Genomic Map